Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-ethynylpyrimidine-2,4(1H,3H)-dione 95%

Supplier: AMBEED, INC
Warning

A124323-1G A124323-100MG A124323-1MG A124323-250MG A124323-1G
103799-316EA 45 USD InStock
103799-316 103800-516 77161-512 77161-514 77161-516 77312-654
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-ethynylpyrimidine-2,4(1H,3H)-dione 95%
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-ethynylpyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00876719
CAS Number: 69075-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty