Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-1H-indazole 98%

Supplier: AMBEED, INC
Warning

A167992-1G A167992-5G A167992-1G A167992-250MG A167992-5G
103806-240EA 27 USD InStock
103806-240 103844-498 77081-536 77163-386 77300-916
5-Fluoro-1H-indazole 98%
5-Fluoro-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₅FN₂
Boiling Pt: 274.9 °C (760 mm Hg)
Melting Pt: 119…125 °C
Flash Pt: 120.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04972877
CAS Number: 348-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty