Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-1H-indole-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A188921-1G A188921-5G A188921-100MG A188921-250MG A188921-5G
103816-142EA 23 USD InStock
103816-142 103846-736 76769-818 76769-820 77084-484
5-Fluoro-1H-indole-3-carboxylic acid 98%
5-Fluoro-1H-indole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₆FNO₂
Boiling Pt: 422.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00047171
CAS Number: 23077-43-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty