Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-1-methyl-3-nitropyridin-2(1H)-one 95%

Supplier: AMBEED, INC
Warning

A457985-5G A457985-1G A457985-250MG
103827-878EA 157 USD InStock
103827-878 103838-954 76842-822 77168-914
5-Fluoro-1-methyl-3-nitropyridin-2(1H)-one 95%
5-Fluoro-1-methyl-3-nitropyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₆H₅FN₂O₃
Boiling Pt: 222.2±40.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD28099326
CAS Number: 1616526-85-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty