Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-methoxyphenol 98%

Supplier: AMBEED, INC
Warning

A874154-100G A874154-10G A874154-1G A874154-25G A874154-5G
103841-294EA 14 USD InStock
103841-294 76820-820 76820-822 76845-558 76845-560 76845-562
5-Fluoro-2-methoxyphenol 98%
5-Fluoro-2-methoxyphenol
Precaución: for Research Use Only


Formula: C₇H₇FO₂
Boiling Pt: 223 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00143459
CAS Number: 72955-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty