Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-methylbenzoxazole 95%

Supplier: AMBEED, INC
Warning

A135403-1G A135403-250MG A135403-1G A135403-5G
103802-148EA 22 USD InStock
103802-148 77095-182 77314-962 77314-964
5-Fluoro-2-methylbenzoxazole 95%
5-Fluoro-2-methylbenzoxazole
Precaución: for Research Use Only


Formula: C₈H₆FNO
MW: 151.14 g/mol
Boiling Pt: 205.9 °C (760 mm Hg)
Flash Pt: 78.3 °C
Storage Temperature: Ambient
MDL Number: MFCD02093471
CAS Number: 701-16-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty