Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-methylindoline 95%

Supplier: AMBEED, INC
Warning

A170563-1G A170563-5G A170563-100MG A170563-250MG A170563-1G
103806-514EA 110 USD InStock
103806-514 103844-788 76824-482 76824-484 77301-064
5-Fluoro-2-methylindoline 95%
5-Fluoro-2-methylindoline
Precaución: for Research Use Only


Formula: C₉H₁₀FN
Storage Temperature: Refrigerator
MDL Number: MFCD19300982
CAS Number: 825-70-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty