Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-3-methoxy-2-nitrobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A518116-1G A518116-5G A518116-100MG A518116-250MG A518116-5G
103828-484EA 104 USD InStock
103828-484 103839-588 76991-432 76991-434 77307-698
5-Fluoro-3-methoxy-2-nitrobenzoic acid 95%
5-Fluoro-3-methoxy-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆FNO₅
MW: 215.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22565642
CAS Number: 1007113-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty