Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluororesorcinol 95%

Supplier: AMBEED, INC
Warning

A137574-1G A137574-100MG A137574-250MG A137574-5G A137574-1G
103802-474EA 49 USD InStock
103802-474 76984-048 76984-050 76984-052 77298-910
5-Fluororesorcinol 95%
5-Fluororesorcinol
Precaución: for Research Use Only


Formula: C₆H₅FO₂
MW: 128.10 g/mol
Boiling Pt: 237.878 °C (760 mm Hg)
Flash Pt: 97.665 °C
Storage Temperature: Ambient
MDL Number: MFCD08457182
CAS Number: 75996-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty