Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluorobenzo[d]thiazole 97%

Supplier: AMBEED, INC
Warning

A160883-1G A160883-5G A160883-250MG A160883-1G
103805-430EA 78 USD InStock
103805-430 103843-666 76768-558 77300-502
5-Fluorobenzo[d]thiazole 97%
5-Fluorobenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₇H₄FNS
Storage Temperature: Refrigerator
MDL Number: MFCD06659658
CAS Number: 1644-85-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty