Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluorooxindole 98%

Supplier: AMBEED, INC
Warning

A228764-5G A228764-25G A228764-10G A228764-500G A228764-5G A228764-100G
103850-322EA 11 USD InStock
103850-322 77089-124 77275-720 77275-722 77303-834 77748-386
5-Fluorooxindole 98%
5-Fluorooxindole
Precaución: for Research Use Only


Formula: C₈H₆FNO
Boiling Pt: 307.2 °C (760 mm Hg)
Melting Pt: 143…147 °C
Flash Pt: 139.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02179598
CAS Number: 56341-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty