Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroisatin 98%

Supplier: AMBEED, INC
Warning

A180513-5G A180513-25G A180513-100G A180513-100MG A180513-10G A180513-500G A180513-5G
103845-900EA 6 USD InStock
103845-900 77083-396 77274-034 77274-036 77274-038 77274-040 77301-676
5-Fluoroisatin 98%
5-Fluoroisatin
Precaución: for Research Use Only


Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 224…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty