Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroisophthalic acid 97%

Supplier: AMBEED, INC
Warning

A275590-1G A275590-5G A275590-250MG A275590-1G A275590-25G A275590-5G
103813-266EA 34 USD InStock
103813-266 103853-190 77238-810 77305-202 77305-204 77305-206
5-Fluoroisophthalic acid 97%
5-Fluoroisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅FO₄
MW: 184.121 g/mol
Boiling Pt: 407.7 °C (760 mm Hg)
Flash Pt: 200.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00230551
CAS Number: 1583-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty