Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-6-(methylamino)pyridine 95%

Supplier: AMBEED, INC
Warning

A101888-1G A101888-5G A101888-100MG A101888-10G A101888-250MG A101888-1G A101888-5G
103796-198EA 59 USD InStock
103796-198 103821-892 77160-220 77160-222 77160-224 77308-780 77308-782
3-Fluoro-6-(methylamino)pyridine 95%
3-Fluoro-6-(methylamino)pyridine
Precaución: for Research Use Only


Formula: C₆H₇FN₂
MW: 126.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848519
CAS Number: 868636-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty