Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formylsalicylic acid 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A235244-5G A235244-100G A235244-10G A235244-500G A235244-25G A235244-5G
103850-762EA 5 USD InStock
103850-762 76815-264 76815-266 76815-268 77089-766 77089-768
5-Formylsalicylic acid 98% (by HPLC)
5-Formylsalicylic acid
Precaución: for Research Use Only


Formula: C₈H₆O₄
Melting Pt: 250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006945
CAS Number: 616-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty