Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydrazinyl-2-methoxypyridine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A107627-1G A107627-5G A107627-100MG A107627-250MG A107627-1G
103797-220EA 69 USD InStock
103797-220 103822-986 76821-636 76821-638 77309-846
5-Hydrazinyl-2-methoxypyridine hydrochloride 97%
5-Hydrazinyl-2-methoxypyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₀ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD23701461
CAS Number: 179543-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty