Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-2-methoxylpyridine 98%

Supplier: AMBEED, INC
Warning

A142359-1G A142359-100MG A142359-10G A142359-1G A142359-5G
103803-150EA 12 USD InStock
103803-150 77109-872 77109-874 77316-376 77316-378
5-Hydroxy-2-methoxylpyridine 98%
5-Hydroxy-2-methoxylpyridine
Precaución: for Research Use Only


Formula: C6H7NO2
Boiling Pt: 330.1 °C (760 mm Hg)
Flash Pt: 153.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06858737
CAS Number: 51834-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty