Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxyisoindolin-1-one 97%

Supplier: AMBEED, INC
Warning

A192832-1G A192832-5G A192832-100MG A192832-250MG A192832-5G
103816-482EA 46 USD InStock
103816-482 103847-092 76974-926 76974-928 77084-974
5-Hydroxyisoindolin-1-one 97%
5-Hydroxyisoindolin-1-one
Precaución: for Research Use Only


Formula: C8H7NO2
Boiling Pt: 519.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10000830
CAS Number: 252061-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty