Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxyisophthalonitrile 97%

Supplier: AMBEED, INC
Warning

A112341-1G A112341-5G A112341-10G A112341-250MG A112341-1G A112341-5G
103798-002EA 30 USD InStock
103798-002 103823-836 77025-840 77025-842 77310-642 77310-644
5-Hydroxyisophthalonitrile 97%
5-Hydroxyisophthalonitrile
Precaución: for Research Use Only


Formula: C8H4N2O
Storage Temperature: Ambient
MDL Number: MFCD11865326
CAS Number: 79370-78-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty