Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxymethylcytosine 95%

Supplier: AMBEED, INC
Warning

A112964-1G A112964-100MG A112964-250MG A112964-1G
103798-138EA 216 USD InStock
103798-138 103799-284 103823-972 77009-020 77009-022 77297-216
5-Hydroxymethylcytosine 95%
5-Hydroxymethylcytosine
Precaución: for Research Use Only


Formula: C₅H₇N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00047369
CAS Number: 1123-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty