Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-2-methylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A162384-1G A162384-5G A162384-100G A162384-10G A162384-25G A162384-1G A162384-5G
103805-608EA 8 USD InStock
103805-608 103843-858 76812-826 76812-828 77300-590 77319-892 77319-894
5-Iodo-2-methylbenzonitrile 97%
5-Iodo-2-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆IN
Boiling Pt: 275.8 °C (760 mm Hg)
Melting Pt: 72…74 °C
Flash Pt: 120.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00128256
CAS Number: 52107-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty