Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-2-methylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A212839-1G A212839-5G A212839-100G A212839-10G A212839-5G
103818-246EA 19 USD InStock
103818-246 103848-942 77249-146 77249-148 77303-264
5-Methoxy-2-methylbenzoic acid 98%
5-Methoxy-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C9H10O3
MW: 166.1739 g/mol
Boiling Pt: 306 °C (760 mm Hg)
Flash Pt: 124.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01846081
CAS Number: 3168-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty