Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl hydrogen glutarate 97%

Supplier: AMBEED, INC
Warning

A159778-5G A159778-25G A159778-5G A159778-500G
103843-510EA 10 USD InStock
103843-510 77300-416 77319-416 77879-094
Methyl hydrogen glutarate 97%
Methyl hydrogen glutarate
Precaución: for Research Use Only


Formula: C₆H₁₀O₄
Boiling Pt: 393.4 °C (760 mm Hg)
Flash Pt: 137.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004409
CAS Number: 1501-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty