Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-7-methyl-1H-indole 95%

Supplier: AMBEED, INC
Warning

A777717-10G A777717-250MG A777717-1G A777717-5G
103840-962EA 109 USD InStock
103840-962 76776-394 76776-396 76845-224 76845-226
5-Methoxy-7-methyl-1H-indole 95%
5-Methoxy-7-methyl-1H-indole
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MW: 161.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027022
CAS Number: 61019-05-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty