Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxyisatin 95%

Supplier: AMBEED, INC
Warning

A332971-1G A332971-25G A332971-5G A332971-100G A332971-10G
103825-622EA 5 USD InStock
103825-622 103836-586 76839-586 76839-588 76839-590 77263-632 77263-634
5-Methoxyisatin 95%
5-Methoxyisatin
Precaución: for Research Use Only


Formula: C₉H₇NO₃
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00169023
CAS Number: 39755-95-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty