Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-quinolyl ether 98%

Supplier: AMBEED, INC
Warning

A751155-5G A751155-1G A751155-10G
103840-852EA 72 USD InStock
103840-852 77308-252 77670-026
Methyl-5-quinolyl ether 98%
Methyl 5-quinolyl ether
Precaución: for Research Use Only


Formula: C₁₀H₉NO
Boiling Pt: 280.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02752664
CAS Number: 6931-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty