Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-γ-Valerolactone 98%

Supplier: AMBEED, INC
Warning

A119285-1000G A119285-100G A119285-25G A119285-1000G A119285-500G
103808-202EA 51 USD InStock
103808-202 76797-274 76797-276 77311-866 77599-720
(±)-γ-Valerolactone 98%
(±)-γ-Valerolactone
Precaución: for Research Use Only


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 207
Melting Pt: -31
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005400
CAS Number: 108-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty