Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylpyrazine-2-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A578021-1G A578021-5G A578021-25G A578021-1G A578021-5G A578021-250MG
103828-846EA 48 USD InStock
103828-846 103839-996 76775-040 77307-858 77307-860 77878-376
5-Methylpyrazine-2-carbonitrile 95%
5-Methylpyrazine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₆H₅N₃
Boiling Pt: 247.1 °C (760 mm Hg)
Flash Pt: 96.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04116510
CAS Number: 98006-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty