Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Morpholinopicolinic acid 95%

Supplier: AMBEED, INC
Warning

A147194-1G A147194-100MG A147194-250MG A147194-50MG A147194-1G
103803-748EA 95 USD InStock
103803-748 76984-502 76984-504 76984-506 77299-572
5-Morpholinopicolinic acid 95%
5-Morpholinopicolinic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₂O₃
Boiling Pt: 446.1±45.0 °C (760 mm Hg)
Flash Pt: 223.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18251373
CAS Number: 1072103-29-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty