Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-3-pyrazolecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A165614-1G A165614-5G A165614-10G A165614-25G A165614-1G A165614-5G A165614-100G
103805-976EA 19 USD InStock
103805-976 103844-230 76768-798 76768-800 77320-444 77320-446 77877-085
5-Nitro-3-pyrazolecarboxylic acid 98%
5-Nitro-3-pyrazolecarboxylic acid
Precaución: for Research Use Only


Formula: C4H3N3O4
MW: 157.0800 g/mol
Boiling Pt: 509.6 °C (760 mm Hg)
Melting Pt: 188…190 °C
Flash Pt: 262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192356
CAS Number: 198348-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty