Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitroisoindolin-1-one 98%

Supplier: AMBEED, INC
Warning

A231062-1G A231062-5G A231062-10G A231062-250MG
103810-750EA 79 USD InStock
103810-750 103850-496 76976-156 77881-598
5-Nitroisoindolin-1-one 98%
5-Nitroisoindolin-1-one
Precaución: for Research Use Only


Formula: C₈H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD10000827
CAS Number: 876343-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty