Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenyl-3-isoxazolecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A114988-1G A114988-10G A114988-25G A114988-1G
103798-514EA 20 USD InStock
103798-514 103824-356 77009-132 77009-134 77297-340
5-Phenyl-3-isoxazolecarboxylic acid 97%
5-Phenyl-3-isoxazolecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇NO₃
Melting Pt: 160…164 °C
Flash Pt: 216.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00464221
CAS Number: 14441-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty