Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Hydroxymethyl)pyridin-3-ol 95%

Supplier: AMBEED, INC
Warning

A203469-1G A203469-5G A203469-100MG A203469-250MG A203469-1G A203469-5G
103817-468EA 233 USD InStock
103817-468 103848-124 76975-292 76975-294 77302-854 77302-856
6-(Hydroxymethyl)pyridin-3-ol 95%
6-(Hydroxymethyl)pyridin-3-ol
Precaución: for Research Use Only


Formula: C₆H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD16249568
CAS Number: 40222-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty