Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Methylamino)nicotinonitrile 98%

Supplier: AMBEED, INC
Warning

A371001-1G A371001-5G A371001-250MG
103826-468EA 29 USD InStock
103826-468 103837-484 76840-728 77003-498
6-(Methylamino)nicotinonitrile 98%
6-(Methylamino)nicotinonitrile
Precaución: for Research Use Only


Formula: C₇H₇N₃
Boiling Pt: 283.5 °C (760 mm Hg)
Melting Pt: 131…133 °C at N/A mmHg
Flash Pt: 125.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689756
CAS Number: 261715-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty