Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-(trifluoromethyl)benzothiazole 98%

Supplier: AMBEED, INC
Danger

A619089-5G A619089-10G A619089-250MG
103840-202EA 47 USD InStock
103840-202 76844-374 77242-774 77242-776
2-Amino-6-(trifluoromethyl)benzothiazole 98%
2-Amino-6-(trifluoromethyl)benzothiazole
Precaución: for Research Use Only


Formula: C₈H₅F₃N₂S
Boiling Pt: 306.7 °C (760 mm Hg)
Melting Pt: 120…124 °C (760 mm Hg)
Flash Pt: 139.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269597
CAS Number: 777-12-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty