Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-2-(trifluoromethyl)pyridine 97%

Supplier: AMBEED, INC
Warning

A100491-1G A100491-5G A100491-250MG A100491-1G A100491-10G
103795-474EA 17 USD InStock
103795-474 103821-168 76765-108 77296-718 77876-779
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
5-Hydroxy-2-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₄F₃NO
MW: 163.1
Boiling Pt: 286.7 °C (760 mm Hg)
Flash Pt: 127.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06801325
CAS Number: 216766-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty