Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dihydro-4(5H)-benzofuranone 98%

Supplier: AMBEED, INC
Warning

A455005-1G A455005-5G A455005-1G A455005-10G A455005-250MG
103827-830EA 7 USD InStock
103827-830 103838-900 76842-748 77252-852 77252-854
6,7-Dihydro-4(5H)-benzofuranone 98%
6,7-Dihydro-4(5H)-benzofuranone
Precaución: for Research Use Only


Formula: C₈H₈O₂
Melting Pt: 30…34 °C
Storage Temperature: Freezer
MDL Number: MFCD00215511
CAS Number: 16806-93-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty