Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-2-cyanobenzothiazole 95%

Supplier: AMBEED, INC
Warning

A233647-1G A233647-100MG A233647-250MG A233647-1G
103810-896EA 371 USD InStock
103810-896 76786-352 76786-354 77303-964
6-Amino-2-cyanobenzothiazole 95%
6-Amino-2-cyanobenzothiazole
Precaución: for Research Use Only


Formula: C₈H₅N₃S
Boiling Pt: 384.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11040279
CAS Number: 7724-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty