Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one 97%

Supplier: AMBEED, INC
Warning

A107071-1G A107071-5G A107071-10G A107071-250MG A107071-5G
103797-118EA 61 USD InStock
103797-118 103822-876 76970-474 76970-476 77309-748
6-Amino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one 97%
6-Amino-3-methyl-2-(methylthio)pyrimidin-4(3H)-one
Precaución: for Research Use Only


Formula: C₆H₉N₃OS
Boiling Pt: 282.3 °C (760 mm Hg)
Melting Pt: 259…261 °C
Flash Pt: 124.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00235002
CAS Number: 54030-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty