Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-5-chloropicolinic acid 97%

Supplier: AMBEED, INC
Warning

A573645-1G A573645-100MG A573645-250MG
103828-818EA 674 USD InStock
103828-818 103839-960 76844-090 77242-282 77242-284
6-Amino-5-chloropicolinic acid 97%
6-Amino-5-chloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18253537
CAS Number: 1256810-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty