Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminopyridazin-3(2H)-one 97%

Supplier: AMBEED, INC
Warning

A226927-1G A226927-5G A226927-100MG A226927-250MG A226927-1G A226927-25G
103819-278EA 51 USD InStock
103819-278 103850-186 76771-022 76771-024 77303-782 77303-784
6-Aminopyridazin-3(2H)-one 97%
6-Aminopyridazin-3(2H)-one
Precaución: for Research Use Only


Formula: C₄H₅N₃O
Boiling Pt: 475.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11520599
CAS Number: 57041-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty