Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminopyridin-2(1H)-one 98%

Supplier: AMBEED, INC
Warning

A107011-1G A107011-5G A107011-100G A107011-25G A107011-1G A107011-5G
103797-112EA 10 USD InStock
103797-112 103822-870 77283-534 77283-536 77309-738 77309-740
6-Aminopyridin-2(1H)-one 98%
6-Aminopyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₅H₆N₂O
Boiling Pt: 358.7 °C (760 mm Hg)
Melting Pt: 211…212 °C
Flash Pt: 170.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08694602
CAS Number: 5154-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty