Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminopyridine-2-sulfonamide 95%

Supplier: AMBEED, INC
Warning

A322579-5G A322579-1G A322579-250MG
103825-306EA 222 USD InStock
103825-306 103836-240 76839-162 77167-336
6-Aminopyridine-2-sulfonamide 95%
6-Aminopyridine-2-sulfonamide
Precaución: for Research Use Only


Formula: C₅H₇N₃O₂S
MW: 173.19 g/mol
Boiling Pt: 455.2±55.0 °C (760 mm Hg)
Melting Pt: 66…168 °C
Storage Temperature: Ambient
MDL Number: MFCD18816592
CAS Number: 75903-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty