Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1,2,3,4-tetrahydroquinoline 98%

Supplier: AMBEED, INC
Warning

A151908-1G A151908-5G A151908-10G A151908-250MG A151908-1G A151908-25G A151908-5G
103804-316EA 10 USD InStock
103804-316 103842-466 77110-336 77110-338 77299-906 77299-908 77299-910
6-Bromo-1,2,3,4-tetrahydroquinoline 98%
6-Bromo-1,2,3,4-tetrahydroquinoline
Precaución: for Research Use Only


Formula: C₉H₁₀BrN
MW: 212.0864 g/mol
Boiling Pt: 303.1 °C (760 mm Hg)
Melting Pt: 32…36 °C
Flash Pt: 137.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04966989
CAS Number: 22190-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty