Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1,2-benzisoxazole 98%

Supplier: AMBEED, INC
Warning

A455704-1G A455704-5G A455704-5G A455704-250MG
103827-842EA 93 USD InStock
103827-842 103838-916 77307-392 77673-144
6-Bromo-1,2-benzisoxazole 98%
6-Bromo-1,2-benzisoxazole
Precaución: for Research Use Only


Formula: C₇H₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD06659634
CAS Number: 1060802-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty