Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoindole-3-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A133135-1G A133135-100G A133135-10G A133135-1G A133135-25G A133135-5G A133135-250MG
103801-832EA 25 USD InStock
103801-832 76996-498 76996-500 77314-442 77314-444 77314-446 77881-055
6-Bromoindole-3-carboxaldehyde 97%
6-Bromoindole-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₉H₆BrNO
Boiling Pt: 235.4 °C (760 mm Hg)
Melting Pt: 202…203 °C
Flash Pt: 88.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00792689
CAS Number: 17826-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty