Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1-methyl-1H-indole 97%

Supplier: AMBEED, INC
Warning

A116020-1G A116020-250MG A116020-1G A116020-5G
103798-660EA 19 USD InStock
103798-660 103824-524 77009-170 77311-346 77311-348
6-Bromo-1-methyl-1H-indole 97%
6-Bromo-1-methyl-1H-indole
Precaución: for Research Use Only


Formula: C₉H₈BrN
MW: 210.08 g/mol
Boiling Pt: 300.878 °C (760 mm Hg)
Flash Pt: 135.766 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08543444
CAS Number: 125872-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty