Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoropyridin-3-ol 95%

Supplier: AMBEED, INC
Warning

A125068-1G A125068-100MG A125068-250MG A125068-1G
103800-620EA 115 USD InStock
103800-620 76781-934 76781-936 77298-032
6-Bromo-2-fluoropyridin-3-ol 95%
6-Bromo-2-fluoropyridin-3-ol
Precaución: for Research Use Only


Formula: C₅H₃BrFNO
MW: 191.986 g/mol
Boiling Pt: 355 °C (760 mm Hg)
Flash Pt: 168.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09999156
CAS Number: 850142-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty