Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-nitropyridin-3-ol 97%

Supplier: AMBEED, INC
Warning

A163990-1G A163990-5G A163990-10G A163990-1G A163990-25G A163990-5G A163990-100G
103805-798EA 10 USD InStock
103805-798 103844-064 76973-692 76973-694 77320-192 77320-194 77320-196 77671-310
6-Bromo-2-nitropyridin-3-ol 97%
6-Bromo-2-nitropyridin-3-ol
Precaución: for Research Use Only


Formula: C₅H₃BrN₂O₃
MW: 218.9900 g/mol
Boiling Pt: 413 °C (760 mm Hg)
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09261204
CAS Number: 443956-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty