Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine 97%

Supplier: AMBEED, INC
Warning

A186108-1G A186108-5G A186108-100G A186108-100MG A186108-10G A186108-250MG A186108-1G A186108-25G A186108-5G
103815-914EA 157 USD InStock
103815-914 103846-488 76872-896 76872-898 76872-900 76872-902 77084-136 77084-138 77302-022
6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine 97%
6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₈H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD17926335
CAS Number: 1207557-36-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty